Potassium (1-(tert-butoxycarbonyl)piperidin-4-yl)trifluoroborate 98%

Supplier: Ambeed
Warning

A607596-1G A607596-250MG A607596-5G
77170-178EA 85 USD
77170-178 77170-180 77170-182
Potassium (1-(tert-butoxycarbonyl)piperidin-4-yl)trifluoroborate 98%
Potassium (1-(tert-butoxycarbonyl)piperidin-4-yl)trifluoroborate

Precaución: for Research Use Only

Formula: C₁₀H₁₈BF₃KNO₂
MW: 291.16 g/mol
MDL Number: MFCD18711957
CAS Number: 1430219-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR