(5-Chloro-2-methylphenyl)methanol 97%

Supplier: Ambeed
Warning

A607021-100MG A607021-1G A607021-250MG A607021-5G
77170-166EA 26.78 USD
77170-166 77170-168 77170-170 77170-172
(5-Chloro-2-methylphenyl)methanol 97%
(5-Chloro-2-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₉ClO
MW: 156.609 g/mol
MDL Number: MFCD11111046
CAS Number: 58966-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR