Cyclopropylacetic acid 97%

Supplier: Ambeed
Danger

A606905-10G A606905-5G A606905-1G
77170-162EA 54.59 USD
77170-162 77170-164 77613-882
Cyclopropylacetic acid 97%
Cyclopropylacetic acid

Precaución: for Research Use Only

Formula: C₅H₈O₂
MW: 100.12 g/mol
MDL Number: MFCD00041544
CAS Number: 5239-82-7
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR