(2-Chloro-phenyl)-oxo-acetonitrile 95%

Supplier: Ambeed
Danger

Synonyms: 2-Chlorobenzoyl cyanide

A606235-1G A606235-250MG
77170-148EA 69.01 USD
77170-148 77170-150
(2-Chloro-phenyl)-oxo-acetonitrile 95%
(2-Chloro-phenyl)-oxo-acetonitrile

Precaución: for Research Use Only

Formula: C₈H₄ClNO
MW: 165.576 g/mol
MDL Number: MFCD03092702
CAS Number: 35022-42-5
UN: 3439
ADR: 6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR