tert-Butyl 4-(dimethylcarbamoyl)piperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A605480-100MG A605480-1G A605480-250MG
77198-050EA 76 USD
77198-050 77198-052 77198-054
tert-Butyl 4-(dimethylcarbamoyl)piperidine-1-carboxylate 98%
tert-Butyl 4-(dimethylcarbamoyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₄N₂O₃
MDL Number: MFCD07776586
CAS Number: 254905-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR