2,4-Dibromo-3-chloropyridine 95%

Supplier: Ambeed
Danger

A604480-100MG A604480-1G A604480-250MG
77170-142EA 35.02 USD
77170-142 77170-144 77170-146
2,4-Dibromo-3-chloropyridine 95%
2,4-Dibromo-3-chloropyridine

Precaución: for Research Use Only

Formula: C₅H₂Br₂ClN
MW: 271.33 g/mol
MDL Number: MFCD00233992
CAS Number: 861024-77-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR