(2,3-Dichloro-4-isobutoxyphenyl)boronic acid 97%

Supplier: Ambeed
Warning

A604455-1G A604455-250MG
77198-046EA 101 USD
77198-046 77198-048
(2,3-Dichloro-4-isobutoxyphenyl)boronic acid 97%
(2,3-Dichloro-4-isobutoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BCl₂O₃
MDL Number: MFCD21609529
CAS Number: 2096329-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR