(2S,4R)-1-Benzyl 2-methyl 4-((tert-butoxycarbonyl)amino)pyrrolidine-1,2-dicarboxylate 98%
Supplier: Ambeed
|
Warning
|
A603603-1G
A603603-250MG
A603603-5G
77170-136EA
142.14
USD
77170-136
77170-138
77170-140
(2S,4R)-1-Benzyl 2-methyl 4-((tert-butoxycarbonyl)amino)pyrrolidine-1,2-dicarboxylate 98%
(2S,4R)-1-Benzyl 2-methyl 4-((tert-butoxycarbonyl)amino)pyrrolidine-1,2-dicarboxylate
Precaución: for Research Use Only
Formula:
C₁₉H₂₆N₂O₆ MW: 378.41 g/mol |
MDL Number:
MFCD28132928 CAS Number: 189215-90-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...