Dimethyl piperidine-2,3-dicarboxylate 95%

Supplier: Ambeed
Warning

A603197-100MG A603197-1G A603197-250MG
77170-130EA 28 USD
77170-130 77170-132 77170-134
Dimethyl piperidine-2,3-dicarboxylate 95%
Dimethyl piperidine-2,3-dicarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₅NO₄
MDL Number: MFCD12026413
CAS Number: 23580-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR