4-(5-Isopropoxy-2-methyl-4-nitrophenyl)pyridine 97%

Supplier: Ambeed
Warning

A602734-1G A602734-5G
77170-126EA 19 USD
77170-126 77170-128
4-(5-Isopropoxy-2-methyl-4-nitrophenyl)pyridine 97%
4-(5-Isopropoxy-2-methyl-4-nitrophenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₅H₁₆N₂O₃
MDL Number: MFCD26407007
CAS Number: 1032903-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR