1-(Phenylsulfinyl)piperidine 98%
Supplier: Ambeed
|
Warning
|
A602581-1G
A602581-5G
77198-042EA
40.17
USD
77198-042
77747-246
1-(Phenylsulfinyl)piperidine 98%
1-(Phenylsulfinyl)piperidine
Precaución: for Research Use Only
Formula:
C₁₁H₁₅NOS MW: 209.30 g/mol |
MDL Number:
MFCD05155800 CAS Number: 4972-31-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...