2-Chloro-3-(trifluoromethyl)isonicotinaldehyde 97%

Supplier: Ambeed
Warning

A602504-100MG A602504-1G A602504-25MG A602504-50MG
77198-034EA 302.82 USD
77198-034 77198-036 77198-038 77198-040
2-Chloro-3-(trifluoromethyl)isonicotinaldehyde 97%
2-Chloro-3-(trifluoromethyl)isonicotinaldehyde

Precaución: for Research Use Only

Formula: C₇H₃ClF₃NO
MW: 209.55 g/mol
MDL Number: MFCD16610709
CAS Number: 1211539-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR