3-Ethoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

A601443-10G A601443-1G A601443-250MG A601443-25G A601443-5G
77198-014EA 27 USD
77198-014 77198-016 77198-018 77198-020 77198-022
3-Ethoxyphenylboronic acid 98%
3-Ethoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₃
MDL Number: MFCD00274219
CAS Number: 90555-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR