1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
8-Amino-1,4-dioxaspiro[4.5]decane-8-carboxylic Acid
A600742-100MG
A600742-250MG
77198-004EA
29.87
USD
77198-004
77198-006
1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal 98%
1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal
Precaución: for Research Use Only
Formula:
C₉H₁₅NO₄ MW: 201.22 g/mol |
MDL Number:
MFCD03790906 CAS Number: 54621-18-0 UN: 1759 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...