Phenazin-1-amine 95%

Supplier: Ambeed
Warning

A600168-100MG A600168-1G A600168-250MG A600168-5G
77170-110EA 37 USD
77170-110 77170-112 77170-114 77893-212
Phenazin-1-amine 95%
Phenazin-1-amine

Precaución: for Research Use Only

Formula: C₁₂H₉N₃
MDL Number: MFCD00705275
CAS Number: 2876-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR