4-Chloro-N-ethyl-5-(trifluoromethyl)pyrimidin-2-amine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-N-ethyl-5-(trifluoromethyl)pyrimidin-4-amine

A600021-100MG A600021-1G A600021-250MG A600021-50MG
77170-102EA 121.54 USD
77170-102 77170-104 77170-106 77170-108
4-Chloro-N-ethyl-5-(trifluoromethyl)pyrimidin-2-amine 95%
4-Chloro-N-ethyl-5-(trifluoromethyl)pyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₇H₇ClF₃N₃
MW: 225.60 g/mol
MDL Number: MFCD23161983
CAS Number: 1374829-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR