1,2,3,6-Tetrahydrophthalimide 98%

Supplier: Ambeed
Warning

Synonyms: Tetrahydrophthalimide

A599273-100G A599273-10G
77197-994EA 25.75 USD
77197-994 77197-996
1,2,3,6-Tetrahydrophthalimide 98%
1,2,3,6-Tetrahydrophthalimide

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.1626 g/mol
MDL Number: MFCD00042782
CAS Number: 85-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR