(Trimethylsilyl)acetonitrile 95%

Supplier: Ambeed
Danger

A598690-10G A598690-1G A598690-250MG A598690-25G A598690-5G
77197-984EA 87 USD
77197-984 77197-986 77197-988 77197-990 77197-992
(Trimethylsilyl)acetonitrile 95%
(Trimethylsilyl)acetonitrile

Precaución: for Research Use Only

Formula: C₅H₁₁NSi
MDL Number: MFCD00001882
CAS Number: 18293-53-3
UN: 1992
ADR: 3,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR