7-Hydroxy-3-(2-methoxyphenyl)-4H-chromen-4-one 97%

Supplier: Ambeed
Warning

A598476-100MG A598476-1G A598476-250MG
77197-978EA 73.13 USD
77197-978 77197-980 77197-982
7-Hydroxy-3-(2-methoxyphenyl)-4H-chromen-4-one 97%
7-Hydroxy-3-(2-methoxyphenyl)-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₁₆H₁₂O₄
MDL Number: MFCD00205542
CAS Number: 63909-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR