(S)-(+)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethyldi-t-butylphosphine ≥98%, ee 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2S)-1-[(1S)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-(diphenylphosphino)ferrocene
A598364-100MG
A598364-1G
A598364-250MG
A598364-5G
A598364-25G
77170-094EA
11
USD
77170-094
77170-096
77170-098
77170-100
77893-208
(S)-(+)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethyldi-t-butylphosphine ≥98%, ee 99%
(S)-(+)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethyldi-t-butylphosphine
Precaución: for Research Use Only
Formula:
C₃₂H₄₀FeP₂ |
MDL Number:
MFCD01074443 CAS Number: 277306-29-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...