2-Amino-5-iodo-4-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Iodo-4-methyl-pyridin-2-ylamine, 5-Iodo-4-methylpyridin-2-ylamine

A598239-10G A598239-250MG A598239-25G A598239-5G
77170-086EA 87.55 USD
77170-086 77170-088 77170-090 77170-092
2-Amino-5-iodo-4-methylpyridine 98%
2-Amino-5-iodo-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₇IN₂
MW: 234.04 g/mol
MDL Number: MFCD06637727
CAS Number: 356561-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR