3-Amino-4-methylpicolinic acid 97%

Supplier: Ambeed
Warning

A597878-100MG A597878-1G
77197-968EA 20.6 USD
77197-968 77197-970
3-Amino-4-methylpicolinic acid 97%
3-Amino-4-methylpicolinic acid

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
MDL Number: MFCD13174786
CAS Number: 53636-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR