trans-Urocanic acid 98%

Supplier: Ambeed
Warning

Synonyms: (E)-3-(1H-Imidazol-5-yl)acrylic acid, 4-Imidazoleacrylic Acid

A597870-250MG A597870-5G
77170-084EA 33.99 USD
77170-084 77594-818
trans-Urocanic acid 98%
trans-Urocanic acid

Precaución: for Research Use Only

Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
MDL Number: MFCD00005203
CAS Number: 3465-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR