(R)-N-(2-Hydroxyethyl)-N,2-dimethyl-8-((5-methylchroman-4-yl)amino)imidazo[1,2-a]pyridine-6-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: PF 03716556

A597315-10MG A597315-1MG A597315-25MG A597315-50MG
77197-960EA 96.82 USD
77197-960 77197-962 77197-964 77197-966
(R)-N-(2-Hydroxyethyl)-N,2-dimethyl-8-((5-methylchroman-4-yl)amino)imidazo[1,2-a]pyridine-6-carboxamide 98%
(R)-N-(2-Hydroxyethyl)-N,2-dimethyl-8-((5-methylchroman-4-yl)amino)imidazo[1,2-a]pyridine-6-carboxamide

Precaución: for Research Use Only

Formula: C₂₂H₂₆N₄O₃
MW: 394.47 g/mol
MDL Number: MFCD19690947
CAS Number: 928774-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR