4-Methoxy-7h-pyrrolo[2,3-d]pyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxy-7H-pyrrolo[2,3-d]pyrimidine

A595831-100MG A595831-250MG A595831-5G
77170-066EA 33.99 USD
77170-066 77170-068 77170-070
4-Methoxy-7h-pyrrolo[2,3-d]pyrimidine 98%
4-Methoxy-7h-pyrrolo[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₇N₃O
MW: 149.15 g/mol
MDL Number: MFCD00127852
CAS Number: 4786-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR