4-Methyl-2-indoleboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

A595272-1G A595272-250MG A595272-5G
77170-060EA 208 USD
77170-060 77170-062 77170-064
4-Methyl-2-indoleboronic acid pinacol ester 98%
4-Methyl-2-indoleboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₀BNO₂
MDL Number: MFCD11858397
CAS Number: 1072811-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR