(3-(Methoxycarbonyl)-5-methylphenyl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methoxycarbonyl-5-methylphenylboronic acid

A595255-100MG A595255-1G A595255-250MG A595255-5G
77170-054EA 37 USD
77170-054 77170-056 77170-058 77893-198
(3-(Methoxycarbonyl)-5-methylphenyl)boronic acid 98%
(3-(Methoxycarbonyl)-5-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₄
MW: 193.99 g/mol
MDL Number: MFCD09751432
CAS Number: 929626-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR