tert-Butyl 2-(4-nitrophenyl)acetate 96%
Supplier: Ambeed
|
Warning
|
A595237-1G
A595237-5G
77197-934EA
50
USD
77197-934
77197-936
tert-Butyl 2-(4-nitrophenyl)acetate 96%
tert-Butyl 2-(4-nitrophenyl)acetate
Precaución: for Research Use Only
Formula:
C₁₂H₁₅NO₄ |
MDL Number:
MFCD24943244 CAS Number: 29704-38-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...