1,2-Bis(2-iodoethoxy)ethane 98% stabilised

Supplier: Ambeed
Warning

Synonyms: Ethylene Glycol Bis(2-iodoethyl) Ether

Stabilization: Stabilised with copper chip

A594428-10G A594428-5G A594428-1G A594428-25G
77197-930EA 56 USD
77197-930 77197-932 77617-500 77747-229
1,2-Bis(2-iodoethoxy)ethane 98% stabilised
1,2-Bis(2-iodoethoxy)ethane

Precaución: for Research Use Only

Formula: C₆H₁₂I₂O₂
MW: 369.97 g/mol
MDL Number: MFCD00075008
CAS Number: 36839-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR