1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine 98%

Supplier: Ambeed
Warning

Synonyms: DOPE, 1,2-Dioleoyl-sn-glycero-3-PE, L-α-Phosphatidylethanolamine, dioleoyl, 2-Aminoethyl-(R)-2,3-bis(oleoyloxy)propyl hydrogen phosphate, 3-sn-Phosphatidylethanolamine, 1,2-dideoyl, (Z)-(2R)-3-(((2-Aminoethoxy)(hydroxy)phosphoryl)oxy)propane-1,2-diyl dioleate

A593796-1G A593796-250MG A593796-100MG
77197-924EA 304.88 USD
77197-924 77197-926 77617-496
1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine 98%
1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine

Precaución: for Research Use Only

Formula: C₄₁H₇₈NO₈P
MW: 744.03 g/mol
MDL Number: MFCD00057986
CAS Number: 4004-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR