α,α,α',α'-Tetrafluoro-p-xylene ≥98%

Supplier: Ambeed
Danger

A593111-1G A593111-25G A593111-5G
77197-906EA 27 USD
77197-906 77197-908 77197-910
α,α,α',α'-Tetrafluoro-p-xylene ≥98%
α,α,α',α'-Tetrafluoro-p-xylene

Precaución: for Research Use Only

Formula: C₈H₆F₄
MDL Number: MFCD01320708
CAS Number: 369-54-0
UN: 1760
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR