7-Bromo-5-chloro-1H-pyrazolo[4,3-b]pyridine 98%

Supplier: Ambeed
Warning

A590836-100MG A590836-1G A590836-250MG
77197-900EA 267.8 USD
77197-900 77197-902 77197-904
7-Bromo-5-chloro-1H-pyrazolo[4,3-b]pyridine 98%
7-Bromo-5-chloro-1H-pyrazolo[4,3-b]pyridine

Precaución: for Research Use Only

Formula: C₆H₃BrClN₃
MW: 232.47 g/mol
MDL Number: MFCD30185930
CAS Number: 1351813-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR