(S)-1-Cbz-2-Methylpiperazine 98%

Supplier: Ambeed
Warning

A590805-1G A590805-25G A590805-5G
77197-894EA 36 USD
77197-894 77197-896 77197-898
(S)-1-Cbz-2-Methylpiperazine 98%
(S)-1-Cbz-2-Methylpiperazine

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₂
MW: 234.29 g/mol
MDL Number: MFCD03840851
CAS Number: 923565-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR