2-(2-Methyl-5-nitro-1H-imidazol-1-yl)ethanamine dihydrochloride hydrate 95%

Supplier: Ambeed
Danger

A590696-100MG A590696-1G A590696-250MG
77197-888EA 23.69 USD
77197-888 77197-890 77197-892
2-(2-Methyl-5-nitro-1H-imidazol-1-yl)ethanamine dihydrochloride hydrate 95%
2-(2-Methyl-5-nitro-1H-imidazol-1-yl)ethanamine dihydrochloride hydrate

Precaución: for Research Use Only

Formula: C₆H₁₄Cl₂N₄O₃
MW: 261.10 g/mol
MDL Number: MFCD00066965
CAS Number: 2828446-98-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR