1-Boc-3-(aminomethyl)-3-methylazetidine 98%

Supplier: Ambeed
Warning

A590028-250MG A590028-5G
77170-034EA 75.99 USD
77170-034 77170-036
1-Boc-3-(aminomethyl)-3-methylazetidine 98%
1-Boc-3-(aminomethyl)-3-methylazetidine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD12408560
CAS Number: 1158758-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR