(E)-3-Phenylprop-2-en-1-amine hydrochloride 97%

Supplier: Ambeed
Warning

A589417-100MG A589417-1G A589417-250MG A589417-5G
77197-876EA 65 USD
77197-876 77197-878 77197-880 77197-882
(E)-3-Phenylprop-2-en-1-amine hydrochloride 97%
(E)-3-Phenylprop-2-en-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂ClN
MDL Number: MFCD00184831
CAS Number: 17480-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR