Methyl (1R,2S,5S)-6,6-dimethyl-3-azabicyclo[3.1.0]hexane-2-carboxylate hydrochloride 96%

Supplier: Ambeed
Warning

Synonyms: (1R,2S,5S)-Methyl 6,6-dimethyl-3-azabicyclo[3.1.0]hexane-2-carboxylate hydrochloride

A589101-100G A589101-100MG A589101-10G A589101-1G A589101-250MG A589101-25G A589101-5G
77170-012EA 1236 USD
77170-012 77170-014 77170-016 77170-018 77170-020 77170-022 77170-024
Methyl (1R,2S,5S)-6,6-dimethyl-3-azabicyclo[3.1.0]hexane-2-carboxylate hydrochloride 96%
Methyl (1R,2S,5S)-6,6-dimethyl-3-azabicyclo[3.1.0]hexane-2-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₅NO₂·ClH
MW: 205.682 g/mol
MDL Number: MFCD11865137
CAS Number: 565456-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR