1,1,1-Trifluoroacetylacetone 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
1,1,1-Trifluoropentane-2,4-dione, 1,1,1-Trifluoro-2,4-pentanedione
A588927-10G
A588927-25G
A588927-5G
77197-866EA
8
USD
77197-866
77197-868
77197-870
1,1,1-Trifluoroacetylacetone 95%
1,1,1-Trifluoroacetylacetone
Precaución: for Research Use Only
Formula:
C₅H₅F₃O₂ MW: 154.09 g/mol Boiling Pt: 107 Flash Pt: 28 |
MDL Number:
MFCD00000427 CAS Number: 367-57-7 UN: 1224 ADR: 3 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...