2-(2-Fluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane 98%

Supplier: Ambeed
Warning

A588881-1G A588881-250MG A588881-5G
77197-860EA 49 USD
77197-860 77197-862 77197-864
2-(2-Fluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane 98%
2-(2-Fluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane

Precaución: for Research Use Only

Formula: C₁₁H₁₄BFO₂
MDL Number: MFCD06200700
CAS Number: 346656-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR