Ferrocenylmethyl methacrylate ≥98% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ) 0.1-0.2%

A588439-100MG A588439-1G A588439-5G
77170-004EA 42 USD
77170-004 77170-006 77170-008
Ferrocenylmethyl methacrylate ≥98% stabilized
Ferrocenylmethyl methacrylate

Precaución: for Research Use Only

Formula: C₁₅H₇FeO₂
MDL Number: MFCD12546011
CAS Number: 31566-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR