3-Fluoro-4-(4-morpholinylcarbonyl)benzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-Fluoro-4-(morpholine-4-carbonyl)phenyl)boronic acid

A587892-1G A587892-250MG A587892-5G
77169-994EA 119.48 USD
77169-994 77169-996 77169-998
3-Fluoro-4-(4-morpholinylcarbonyl)benzeneboronic acid 98%
3-Fluoro-4-(4-morpholinylcarbonyl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃BFNO₄
MW: 253.04 g/mol
MDL Number: MFCD20265298
CAS Number: 1008119-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR