trans-3-Aminocyclobutanecarboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A586669-250MG
A586669-100MG
A586669-1G
A586669-50MG
77169-988EA
287.37
USD
77169-988
77684-448
77684-449
77684-450
trans-3-Aminocyclobutanecarboxylic acid 97%
trans-3-Aminocyclobutanecarboxylic acid
Precaución: For research use only.
Formula:
C₅H₉NO₂ |
MDL Number:
MFCD06659965 CAS Number: 74307-75-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...