[2-(Benzyloxy)ethoxy]acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: BnO-PEG1-CH2COOH, 2-(2-(Benzyloxy)ethoxy)acetic acid

A586572-1G A586572-250MG A586572-100MG
77169-984EA 186.43 USD
77169-984 77613-816 77684-447
[2-(Benzyloxy)ethoxy]acetic acid 98%
[2-(Benzyloxy)ethoxy]acetic acid

Precaución: For research use only.

Formula: C₁₁H₁₄O₄
MW: 210.226 g/mol
MDL Number: MFCD16294180
CAS Number: 93206-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR