1-Boc-indoline-5-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 5-(tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-indole-1-carboxylate, tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indoline-1-carboxylate

A586227-100MG A586227-1G A586227-250MG A586227-5G
77169-976EA 46.35 USD
77169-976 77169-978 77169-980 77169-982
1-Boc-indoline-5-boronic acid pinacol ester 98%
1-Boc-indoline-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₂₈BNO₄
MW: 345.246 g/mol
MDL Number: MFCD12408237
CAS Number: 837392-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR