1,2-Dimyristoyl-sn-glycero-3-phosphoethanolamine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,2-Dimyristoyl-sn-glycero-3-phosphoethanolamine, (2R)-3-(((2-Aminoethoxy)(hydroxy)phosphoryl)oxy)propane-1,2-diyl ditetradecanoate, L-DMPE, 2-Aminoethyl (R)-2,3-Bis(tetradecanoyloxy)propyl Hydrogen Phosphate, 1,2-Ditetradecanoyl-sn-glycero-3-phosphoethanolamine
A585917-1MG
A585917-250MG
A585917-100MG
77197-844EA
5.15
USD
77197-844
77617-462
77747-212
1,2-Dimyristoyl-sn-glycero-3-phosphoethanolamine 97%
1,2-Dimyristoyl-sn-glycero-3-phosphoethanolamine
Precaución: for Research Use Only
Formula:
C₃₃H₆₆NO₈P MW: 635.86 g/mol |
MDL Number:
MFCD00036775 CAS Number: 998-07-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...