N-(3-Fluoro-4-methoxyphenyl)acetamide 95%

Supplier: Ambeed
Warning

A585267-1G A585267-250MG
77169-966EA 33 USD
77169-966 77169-968
N-(3-Fluoro-4-methoxyphenyl)acetamide 95%
N-(3-Fluoro-4-methoxyphenyl)acetamide

Precaución: for Research Use Only

Formula: C₉H₁₀FNO₂
MDL Number: MFCD12913364
CAS Number: 452-15-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR