4-(4-Methylpiperazin-1-yl)pyridin-2-amine 97%

Supplier: Ambeed
Warning

A583762-1G A583762-250MG
77197-842EA 206 USD
77197-842 77747-209
4-(4-Methylpiperazin-1-yl)pyridin-2-amine 97%
4-(4-Methylpiperazin-1-yl)pyridin-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₄
MW: 192.26 g/mol
MDL Number: MFCD19543946
CAS Number: 959986-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR