2',3',4',5'-Tetrafluoroacetophenone 98%

Supplier: Ambeed
Warning

A583188-1G A583188-25G A583188-5G
77197-836EA 28 USD
77197-836 77197-838 77197-840
2',3',4',5'-Tetrafluoroacetophenone 98%
2',3',4',5'-Tetrafluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₄F₄O
MDL Number: MFCD00134310
CAS Number: 66286-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR