UK-5099 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-cyano-3-(1-phenyl-1H-indol-3-yl)-2-propenoic acid, 2-Cyano-3-(1-phenyl-1H-indol-3-yl)acrylic acid
A582535-100MG
A582535-10MG
A582535-1MG
A582535-25MG
A582535-50MG
A582535-5MG
A582535-1G
A582535-250MG
77169-948EA
118.45
USD
77169-948
77169-950
77169-952
77169-954
77169-956
77169-958
77684-439
77684-440
UK-5099 98%
UK-5099
Precaución: For research use only.
Formula:
C₁₈H₁₂N₂O₂ MW: 288.3 g/mol |
MDL Number:
MFCD19443867 CAS Number: 56396-35-1 UN: 3077 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...