4,5-Diamino-2-(trifluoromethyl)benzonitrile 95+%

Supplier: Ambeed
Warning

A582175-1G A582175-250MG A582175-5G
77169-940EA 74 USD
77169-940 77169-942 77169-944
4,5-Diamino-2-(trifluoromethyl)benzonitrile 95+%
4,5-Diamino-2-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₆F₃N₃
MDL Number: MFCD18074368
CAS Number: 882978-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR